Assay Details
Gene Symbol: MRPL52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495223.6 | XP_001495273.4 | 254 | 105 | ||
XM_005613738.3 | XP_005613795.1 | 261 | 105 | |||
XM_005613739.3 | XP_005613796.1 | 262 | 105 | |||
XM_005613740.3 | XP_005613797.1 | 252 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
