Assay Details
Gene Symbol: NUAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640396.1 | XP_023496164.1 | 817 | 73 | ||
XM_023640397.1 | XP_023496165.1 | 817 | 73 | |||
XM_023640398.1 | XP_023496166.1 | 814 | 73 | |||
XM_023640399.1 | XP_023496167.1 | 2454 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
