Assay Details
Gene Symbol: LEMD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364923.4 | XP_003364971.1 | 481 | 84 | ||
XM_023640405.1 | XP_023496173.1 | 584 | 84 | |||
XM_023640406.1 | XP_023496174.1 | 635 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
