Assay Details
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739419.2 | XP_014594905.2 | 960 | 92 | ||
XM_023640410.1 | XP_023496178.1 | 960 | 92 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
