Assay Details
Gene Symbol: FCMR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640457.1 | XP_023496225.1 | 869 | 59 | ||
XM_023640458.1 | XP_023496226.1 | 869 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
