Assay Details
Gene Symbol: POGK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640469.1 | XP_023496237.1 | 195 | 70 | ||
XM_023640470.1 | XP_023496238.1 | 339 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
