Assay Details
Gene Symbol: PIGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489837.5 | XP_001489887.1 | 1320 | 69 | ||
XM_014728766.2 | XP_014584252.1 | 1190 | 69 | |||
XM_023633267.1 | XP_023489035.1 | 1947 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
