Assay Details
Gene Symbol: AP1S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634135.1 | XP_023489903.1 | 306 | 82 | ||
XM_023634136.1 | XP_023489904.1 | 304 | 82 | |||
XM_023634137.1 | XP_023489905.1 | 306 | 82 | |||
XM_023634138.1 | XP_023489906.1 | 306 | 82 | |||
XM_023634139.1 | XP_023489907.1 | 306 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
