Assay Details
Gene Symbol: PDHA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492157.6 | XP_001492207.2 | 1136 | 76 | ||
GenBank mRNA | JL616099.2 | - | 4 - 5 | 409 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
