Assay Details
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493681.4 | XP_001493731.1 | 634 | 64 | ||
XM_005614114.3 | XP_005614171.1 | 499 | 64 | |||
XM_023633299.1 | XP_023489067.1 | 603 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
