Assay Details
Gene Symbol: HSD17B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495307.5 | XP_001495357.1 | 490 | 77 | ||
XM_014728917.2 | XP_014584403.1 | 493 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
