Assay Details
Gene Symbol: VSIG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614230.3 | XP_005614287.2 | 937 | 67 | ||
XM_005614232.3 | XP_005614289.2 | 938 | 67 | |||
GenBank mRNA | JL622856.1 | - | 4 - 5 | 407 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
