Assay Details
Gene Symbol: PLP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014729058.2 | XP_014584544.1 | 845 | 72 | ||
XM_023634324.1 | XP_023490092.1 | 740 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
