Assay Details
Gene Symbol: RNF128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492036.4 | XP_001492086.2 | 1061 | 66 | ||
XM_003365831.4 | XP_003365879.1 | 975 | 66 | |||
XM_023633276.1 | XP_023489044.1 | 1064 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
