Assay Details
Gene Symbol: BCAP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005614625.3 | XP_005614682.1 | 786 | 62 | ||
XM_023633930.1 | XP_023489698.1 | 805 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
