Assay Details
Gene Symbol: ABCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491684.4 | XP_001491734.1 | 2059 | 65 | ||
XM_023633929.1 | XP_023489697.1 | 2193 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
