Assay Details
Gene Symbol: RPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014729298.2 | XP_014584784.1 | 154 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014729298.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
