Assay Details
Gene Symbol: BRCC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498791.6 | XP_001498841.2 | 474 | 68 | ||
XM_003365914.4 | XP_003365962.1 | 471 | 68 | |||
XM_023634086.1 | XP_023489854.1 | 422 | 68 | |||
XM_023634087.1 | XP_023489855.1 | 404 | 68 | |||
GenBank mRNA | JL615923.1 | - | 5 - 6 | 406 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
