Assay Details
Gene Symbol: CLIC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494262.5 | XP_001494312.2 | 968 | 72 | ||
GenBank mRNA | JL618287.1 | - | 2 - 3 | 294 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
