Assay Details
Gene Symbol: TMLHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498280.5 | XP_001498330.1 | 799 | 69 | ||
XM_005614678.3 | XP_005614735.1 | 800 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
