Assay Details
Gene Symbol: VPS29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495080.6 | XP_001495130.1 | 312 | 78 | ||
XM_001495099.6 | XP_001495149.1 | 329 | 78 | |||
GenBank mRNA | JL615882.2 | - | 1 - 2 | 189 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
