Assay Details
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005610077.3 | XP_005610134.2 | 830 | 81 | ||
XM_005610078.3 | XP_005610135.2 | 848 | 81 | |||
XM_014739808.2 | XP_014595294.1 | 890 | 81 | |||
XM_014739809.2 | XP_014595295.1 | 703 | 81 | |||
GenBank mRNA | JL616170.1 | - | 5 - 6 | 686 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
