Assay Details
Gene Symbol: IKBIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631278.1 | XP_023487046.1 | 2859 | 114 | ||
GenBank mRNA | JL621659.1 | - | 1 | 614 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
