Assay Details
Gene Symbol: BBIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005602236.3 | XP_005602293.1 | 120 | 85 | ||
XM_023639544.1 | XP_023495312.1 | 477 | 85 | |||
XM_023639548.1 | XP_023495316.1 | 134 | 85 | |||
XM_023639553.1 | XP_023495321.1 | 505 | 85 | |||
XM_023639554.1 | XP_023495322.1 | 230 | 85 | |||
XM_023639556.1 | XP_023495324.1 | 360 | 85 | |||
XM_023639560.1 | XP_023495328.1 | 386 | 85 | |||
GenBank mRNA | JL621929.1 | - | 2 - 3 | 138 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005602236.3, XM_023639544.1, XM_023639548.1, XM_023639553.1, XM_023639554.1, XM_023639556.1, XM_023639560.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
