Assay Details
Gene Symbol: MPZL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500934.4 | XP_001500984.1 | 2464 | 98 | ||
GenBank mRNA | JL626973.1 | - | 1 | 216 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
