Assay Details
Gene Symbol: SLC25A44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500013.3 | XP_001500063.1 | 2933 | 117 | ||
GenBank mRNA | JL628485.1 | - | 1 | 122 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
