Assay Details
Gene Symbol: IGFBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014736810.2 | XP_014592296.2 | 641 | 94 | ||
GenBank mRNA | AY752933.1 | - | 1 - 2 | 293 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
