Assay Details
Gene Symbol: RPS3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501474.4 | XP_001501524.3 | 662 | 84 | ||
GenBank mRNA | EU687563.1 | - | 4 - 5 | 554 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
