Assay Details
Gene Symbol: NFKBIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001497571.5 | XP_001497621.1 | 158 | 63 | ||
XM_023649683.1 | XP_023505451.1 | 158 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
