Assay Details
Gene Symbol: PRRX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001495718.4 | XP_001495768.1 | 1293 | 85 | ||
XM_023640540.1 | XP_023496308.1 | 1293 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
