Assay Details
Gene Symbol: COP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640593.1 | XP_023496361.1 | 1721 | 67 | ||
XM_023640594.1 | XP_023496362.1 | 1709 | 67 | |||
XM_023640595.1 | XP_023496363.1 | 1721 | 67 | |||
XM_023640596.1 | XP_023496364.1 | 1721 | 67 | |||
XM_023640597.1 | XP_023496365.1 | 1652 | 67 | |||
XM_023640598.1 | XP_023496366.1 | 1721 | 67 | |||
XM_023640599.1 | XP_023496367.1 | 1176 | 67 | |||
XM_023640600.1 | XP_023496368.1 | 1228 | 67 | |||
XM_023640601.1 | XP_023496369.1 | 879 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
