Assay Details
Gene Symbol: MR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005609762.3 | XP_005609819.1 | 428 | 61 | ||
XM_005609764.3 | XP_005609821.1 | 430 | 61 | |||
XM_014739668.2 | XP_014595154.1 | 431 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
