Assay Details
Gene Symbol: ARPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915096.5 | XP_001915131.2 | 588 | 82 | ||
GenBank mRNA | HQ890132.1 | - | 1 - 2 | 171 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
