Assay Details
Gene Symbol: PROX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488456.5 | XP_001488506.1 | 2387 | 79 | ||
XM_005609798.3 | XP_005609855.1 | 2390 | 79 | |||
XM_005609799.3 | XP_005609856.1 | 2497 | 79 | |||
XM_005609800.3 | XP_005609857.1 | 2431 | 79 | |||
XM_023640735.1 | XP_023496503.1 | 2220 | 79 | |||
XM_023640736.1 | XP_023496504.1 | 4017 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
