Assay Details
Gene Symbol: BRCA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621924.1 | XP_023477692.1 | 6670 | 98 | ||
XM_023621925.1 | XP_023477693.1 | 6374 | 98 | |||
XM_023621926.1 | XP_023477694.1 | 6511 | 98 | |||
XM_023621927.1 | XP_023477695.1 | 5927 | 98 | |||
XM_023621928.1 | XP_023477696.1 | 5872 | 98 | |||
XM_023621929.1 | XP_023477697.1 | 6673 | 98 | |||
XM_023621930.1 | XP_023477698.1 | 6673 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
