Assay Details
Gene Symbol: BAG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023628474.1 | XP_023484242.1 | 3170 | 107 | ||
XM_023628475.1 | XP_023484243.1 | 3188 | 107 | |||
XM_023628476.1 | XP_023484244.1 | 3231 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
