Assay Details
Gene Symbol: SYT14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023640779.1 | XP_023496547.1 | 2707 | 100 | ||
XM_023640780.1 | XP_023496548.1 | 2650 | 100 | |||
XM_023640782.1 | XP_023496550.1 | 1672 | 100 | |||
XM_023640783.1 | XP_023496551.1 | 1613 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
