Assay Details
Gene Symbol: ALG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014741234.2 | XP_014596720.1 | 1932 | 64 | ||
XM_023644796.1 | XP_023500564.1 | 1992 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
