Assay Details
Gene Symbol: LPGAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001489450.6 | XP_001489500.3 | 1183 | 85 | ||
XM_023640755.1 | XP_023496523.1 | 1131 | 85 | |||
XM_023640756.1 | XP_023496524.1 | 1124 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
