Assay Details
Gene Symbol: RGS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364967.4 | XP_003365015.1 | 262 | 82 | ||
XM_014739674.2 | XP_014595160.1 | 262 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
