Assay Details
Gene Symbol: VSIG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014739972.2 | XP_014595458.2 | 4355 | 67 | ||
XM_023640911.1 | XP_023496679.1 | 4359 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
