Assay Details
Gene Symbol: SPACA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650332.1 | XP_023506100.1 | 310 | 73 | ||
XM_023650333.1 | XP_023506101.1 | 310 | 73 | |||
XM_023650334.1 | XP_023506102.1 | 310 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
