Assay Details
Gene Symbol: RWDD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023650726.1 | XP_023506494.1 | 514 | 86 | ||
XM_023650727.1 | XP_023506495.1 | 606 | 86 | |||
XM_023650728.1 | XP_023506496.1 | 712 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
