Assay Details
Gene Symbol: WFIKKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005597493.3 | XP_005597550.2 | 317 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005597493.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
