Assay Details
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636020.1 | XP_023491788.1 | 406 | 60 | ||
XM_023636021.1 | XP_023491789.1 | 365 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
