Assay Details
Gene Symbol: FGF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737727.2 | XP_014593213.2 | 3152 | 74 | ||
XM_023636127.1 | XP_023491895.1 | 3220 | 74 | |||
XM_023636128.1 | XP_023491896.1 | 3185 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
