Assay Details
Gene Symbol: CDKN1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023624843.1 | XP_023480611.1 | 580 | 90 | ||
XM_023624844.1 | XP_023480612.1 | 552 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
