Assay Details
Gene Symbol: KCNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023626752.1 | XP_023482520.1 | 1081 | 79 | ||
XM_023626753.1 | XP_023482521.1 | 1226 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
