Assay Details
Gene Symbol: HUNK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023630009.1 | XP_023485777.1 | 620 | 111 | ||
XM_023630010.1 | XP_023485778.1 | 620 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
