Assay Details
Gene Symbol: MPST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606606.3 | XP_005606663.1 | 1411 | 104 | ||
XM_005606608.3 | XP_005606665.1 | 2609 | 104 | |||
XM_014736819.2 | XP_014592305.1 | 736 | 104 | |||
XM_023631284.1 | XP_023487052.1 | 637 | 104 | |||
XM_023631285.1 | XP_023487053.1 | 750 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
